Learn More
TAF(II)135 Mouse, Unlabeled, Clone: 22, BD
Descripción
Tightly associated factors (TAFs) play an essential role in transcriptional activation through their interaction with a variety of activators. TAF[II]135 (a.k.a. TAF[II]130) is a human homologue of Drosophila TAF[II]110, the first TAF shown to have coactivator activity. TAF[II]135 contains multiple glutamine-rich regions, as well as a coactivator domain (CAD). The glutamine-rich regions of TAF[II]135 facilitate interaction with Sp1 and CREB, which leads to enhancement of both Sp1- and CREB-mediated transcription. In addition, TAF[II]135 can potentiate transcriptional stimulation by AF-2 of the retinoic acid, thyroid hormone, and vitamine D3 receptors. However TAF[II]135 does not interact with the AF-2s of the estrogen and retinoid X receptors. Interestingly, TAF[II]135 enhancement of CREB transcriptional activity may be disrupted by expanded polyglutamine (CAG) repeats found in at least eight different neurodegenerative disorders. Thus, TAF[II]135 may have important coactivator activities in many different transcription-regulating cell signaling pathways, and interference of TAF[II]135 activity by CAG repeats may be a common mechanism of specific types of neuropathologies.
Host Species: Mouse
Clone: 22
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human TAF[II]135 aa. 454-565
Immunofluorescence, Western Blotting
Especificaciones
Especificaciones
| Antígeno | TAF(II)135 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | 22 |
| Concentración | 250μg/mL |
| Conjugado | Unconjugated |
| Formulación | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Human TAF[II]135 aa. 454-565 |
| Método de purificación | Affinity Purified |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.