missing translation for 'onlineSavingsMsg'
Learn More

CD8, CD4 and CD3 cocktail , APC, FITC, PE, Clone: SK1, BD

Código de producto. 15819538
Change view
Click to view available options
Cantidad:
50 Tests
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15819538 50 Tests cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15819538

Marca: BD Biosciences 558625

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Monoclonal Antibody

Cocktail Component Clone Isotype

APC anti-Human CD8 SK1 mIgG1

FITC anti-Human CD3 SP34-2 mIgG1

The NHP T Lymphocyte Cocktail is a three-color reagent cocktail designed to identify NHP T lymphocytes by direct immunofluorescence staining with flow cytometric analysis. The SK1 antibody reacts with the hinge-like membrane-proximal domain of the 32-kDa alpha chain of the CD8 differentiation antigen.. The CD8α and β chains (CD8a and CD8b, respectively) form a heterodimer on the surface of most thymocytes and a subpopuplation of mature T-lymphocytes (ie, MHC class I-restricted T cells, including most T suppressor/cytotoxic cells). The L200 antibody reacts with the human form of the 56 kDa transmembrane glycoprotein, CD4, present on the T-helper/inducer subset of normal human donor peripheral blood lymphocytes. L200 antibody also cross-reacts with a subset of CD3-positive peripheral blood lymphocytes, but not monocytes, of both rhesus and cynomolgus macaque monkeys. Cross-reactivity on both lymphocytes and monocytes (weak) of baboon is also observed. The distribution on lymphocytes is similar for both human and monkey, with the majority of CD4-positive lymphocytes being CD8-negative and lacking reactivity with antibodies to B- or NK-cell markers. The SP34-2 antibody reacts with the T-cell receptor-associated CD3 cell-surface antigen found on thymocytes and peripheral T lymphocytes. Clone SP34-2 is a mouse IgG1 isotype monoclonal antibody, descendant of SP34 (mouse IgG3), with the same specificity and reactivity pattern as the parent clone. It cross-reacts with a major subset of peripheral blood lymphocytes, but not monocytes or granulocytes, of baboon, and rhesus, cynomolgus, and pigtail macaque monkeys.

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD8, CD4 and CD3 cocktail
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon SK1
Conjugado FITC/APC/PE
Formulación Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Método de purificación Affinity Purified
Cantidad 50 Tests
Estado normativo RUO
Primario o secundario Primary
Especies diana Baboon, Cynomolgus, Rhesus Monkey
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.