missing translation for 'onlineSavingsMsg'
Learn More

T-Bet Mouse, Unlabeled, Clone: 4B10, BD

Código de producto. 15860899
Change view
Click to view available options
Cantidad:
0.1 mg
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15860899 0.1 mg cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15860899

Marca: BD Biosciences 561262

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The 4B10 monoclonal antibody specifically binds to human and mouse T-bet. T-bet (T-box gene expressed in T cells) is a master regulatory transcription factor that is also known as TBX21 (T-box21) and TBLYM (T-box transcription factor, expressed in lymphocytes). Human (535 amino acids; 58.3 kDa predicted molecular mass) and mouse (530 amino acids; 57.7 kDa) T-bet proteins are encoded by the human TBX21 (chromosome 17) and mouse Tbx21 (chromosome 11) genes. The human and mouse T-bet protein amino acid sequences are 88% homologous. Human and mouse T-bet proteins share a highly conserved (98% homologous amino acid sequences) T-box protein domain that is centrally located and mediates binding to DNA. T-bet is expressed by and activates transcriptional activities within hematopoietic cells including stem cells,NK and NKT cells and subsets of thymocytes, primed/activatedCD4+ T cells, CD8+ T cells and γδ T cells, B cells, and dendritic cells. Interferon-gamma (IFN-γ), interleukin-27 (IL-27), and IL-12 act on peripheral antigen-triggered (TCR-signaling) T cells to increase T-bet expression. With respect to T helper lymphocytes, T-bet directs the differentiation of naοve CD4+ precursor T cells to become Th1-like effector and memory cells. T-bet accomplishes this by activating Th1 genetic programs (including epigenetic modifications) while repressing opposing T helper subset programs. T-bet controls the upregulated expression of the Th1 signature cytokine, IFN-γ, the IL-12Rβ2 subunit and the Runx3 transcription factor and can repress the function of other transcriptional regulators, such as GATA-3 (master regulator of Th2 development) and the expression of other cytokines including IL-2, IL-4 and IL-5.

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno T-Bet
Aplicaciones Western Blot
Clasificación Monoclonal
Clon 4B10
Concentración 0.5mg/mL
Conjugado Unconjugated
Descripción T-box expressed in T cells; TBX21; T-box 21; TBLYM
Formulación Aqueous buffered solution containing ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Mouse T-bet Recombinant Protein
Método de purificación Affinity Purified
Cantidad 0.1 mg
Estado normativo RUO
Primario o secundario Primary
Especies diana Human, Mouse
Contenido y almacenamiento Store undiluted at 4°C
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.