Learn More
CD3, CD6 and CD20 cocktail Mouse, APC, FITC, PE, Clone: L27, 3G8, SP34-2, BD
Descripción
Cocktail Component Clone Isotype
PE anti-Human CD16 3G8 mIgG1
APC anti-Human CD20 L27 mIgG1
The NHP T/B/NK Cocktail is a three-color reagent cocktail designed to identify NHP T, B, and NK lymphocyte populations by direct immunofluorescent staining with flow cytometric analysis. Clone SP34-2 is a mouse IgG1 isotype monoclonal antibody, descendant of SP34 (mouse IgG3), with the same specificity and reactivity pattern as the parent clone. It cross-reacts with a major subset of peripheral blood lymphocytes, but not monocytes or granulocytes, of baboon, and rhesus, cynomolgus, and pigtail macaque monkeys. The monoclonal antibody clone L27 reacts with CD20 found only on B lymphocytes. The monoclonal antibody clone 3G8 reacts with CD16, the 50-65 kDa transmembrane form of IgG Fc (Fc γRIII), a human NK-cell-associated antigen. CD16 is expressed on NK cells as well as macrophages and granulocytes.
Especificaciones
Especificaciones
| Antígeno | CD3/CD6/CD20 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | L27, 3G8, SP34-2 |
| Conjugado | FITC/APC/PE |
| Formulación | Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Método de purificación | Affinity Purified |
| Cantidad | 50 Tests |
| Estado normativo | RUO |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.