missing translation for 'onlineSavingsMsg'
Learn More

Stat3 Mouse, PE, Clone: M59-50, BD

Código de producto. 15860159
Change view
Click to view available options
Cantidad:
50 Tests
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15860159 50 Tests cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15860159

Marca: BD Biosciences 560391

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Stat (Signal transducer and activators of transcription) proteins are critical mediators of the biologic activity of cytokines, including interleukins, interferons, erythropoietin, and growth factors. Ligand-receptor interaction leads to activation of constitutively associated JAK family kinases and subsequent recruitment/activation of Stat proteins by tyrosine phosphorylation. Active Stat proteins then move to the nucleus to promote transcription of cytokine-inducible genes. Stat3 is a 92kDa protein that is activated as a DNA-binding protein through cytokines, such as IL-6, and growth factors, such as EGF. Upon activation, Stat3 dimerizes, translocates to the nucleus and binds DNA response elements, thereby regulating gene expression. It has been reported that Stat3 binds to DNA as a homodimer, but it is also capable of binding as a heterodimer with Stat1. Stat3 is widely expressed and can bind to the sis-inducible element (SIE) site from the c-fos promoter. This site is similar to the GAS element that is present in IFN-γ induced genes. The M59-50 monoclonal antibody recognizes Stat3 (isoform 1) regardless of phosphorylation status. The specificity of this antibody conjugate for flow cytometric analysis was validated by confirming that RNA-mediated interference (RNAi) of the specific protein reduced the staining of the cells (see figure). Furthermore, the capacity of the RNAi to down-regulate the expression of the relevant protein was confirmed by western blot analysis.

Host Species: Mouse
Clone: M59-50
Isotype: IgG1 λ
Species Reactivity: Human
Immunogen: Human Stat3 Recombinant Protein

Intracellular Staining

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Stat3
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon M59-50
Conjugado PE
Descripción STAT3; Acute-phase response factor; APRF; HIES
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Human Stat3 Recombinant Protein
Método de purificación Affinity Purified
Cantidad 50 Tests
Estado normativo RUO
Disciplina de investigación Cell Biology
Primario o secundario Primary
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 λ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.