missing translation for 'onlineSavingsMsg'
Learn More

Stat1 (pS727) Mouse, Alexa Fluor 647, Clone: K51-856, BD

Código de producto. 15889998
Change view
Click to view available options
Cantidad:
50 Tests
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15889998 50 Tests cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15889998

Marca: BD Biosciences 560190

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Stat (Signal transducer and activators of transcription) proteins are critical mediators of the biologic activity of cytokines, including interleukins, interferons, erythropoietin, and growth factors. Ligand-receptor interaction leads to activation of constitutively associated JAK family kinases and subsequent recruitment/activation of Stat proteins by tyrosine phosphorylation. Active Stat proteins then move to the nucleus to promote transcription of cytokine-inducible genes. Seven Stat proteins have been cloned, each of which is differentially expressed and/or activated in a cytokine-specific and cell type-specific manner. Stat1 and Stat2 are components of the ISGF3 (Interferon-Stimulated Gene Factor 3) complex, which is the primary transcription activator induced by the binding of the interferon to a specific cell-surface receptor. Stat1 has two alternatively spliced isoforms, 91kDa Stat1α and 84kDa Stat1β; Stat1α has 38 additional C-terminal amino acids. In response to the binding of IFNα, IFNγ, EGF, PDGF, or CSF-1 to their respective receptors, the Stat1 subunits become tyrosine-phosphorylated at Y701, and the complex is translocated to the nucleus. This results in the formation of an active
complex that includes the DNA-binding p48 subunit. This complex is responsible for modulating the transcription of the interferon-stimulated genes (ISGs). Furthermore, phosphorylation of serine 727 (S727) of Stat1 may occur in response to pathogens, cytokines, UV irradiation, and engagement of T and B cell antigen receptors. This second phosphorylation event is necessary for fully effective activation of transcription and may differentially regulate gene activation by interacting with other regulatory factors, such as p53 in the apoptotic pathway. The K51-856 monoclonal antibody recognizes the phosphorylated S727 in the C-terminal transactivating domain of human Stat1α; this site is not present in Stat1β.

Host Species: Mouse
Clone: K51-856
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Phosphorylated Human Stat1 Peptide

Intracellular Staining

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Stat1 (pS727)
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon K51-856
Conjugado Alexa Fluor 647
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Phosphorylated Human Stat1 Peptide
Método de purificación Affinity Purified
Cantidad 50 Tests
Estado normativo RUO
Disciplina de investigación Cell Biology
Primario o secundario Primary
Especies diana Human, Mouse
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.