Learn More
Stat 6 (pY641) Mouse anti-Human, Unlabeled, Clone: 18/P-STAT6, BD
Mouse Monoclonal Antibody
277.00€ - 505.00€
Especificaciones
| Antígeno | Stat 6 (pY641) |
|---|---|
| Clon | 18/P-STAT6 |
| Concentración | 250μg/mL |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|
15835979
|
BD Biosciences
611566 |
50 μg |
277.00€
1 set |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
|
15845979
|
BD Biosciences
611567 |
150 μg |
505.00€
1 set |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
Interleukin-4 (IL-4), a major immunoregulatory cytokine, is secreted by activated T lymphocytes, basophils, and mast cells and plays an important role in modulating T helper cell lineage development. It induces specific gene expression via the tyrosine phosphorylation of Stat6 at tyrosine 641 (Y641). Stat6, a member of the signal transducers and activators of transcription protein family, mediates signals for IL-4 and, possibly, IL-13. While Stat6 is widely expressed in human tissues, it exhibits elevated expression in peripheral blood lymphocytes, colon,
intestine, ovary, prostate, thymus, spleen, kidney, liver, lung, and placenta. Following cytokine receptor ligation, Jak kinases are activated and phosphorylate the cytoplasmic tails of the oligomerized receptors. The SH3:SH2 domain of Stat6 associates with tyrosine-phosphorylated IL-4 receptor and the proximal Jak kinase phosphorylates Stat6 at Y641 on the C-terminal side of the SH2 domain. Stat6 is then released from the receptor, dimerizes, and is thought to contact the basal transcription machinery by binding to p300/CBP. Thus, Stat6 mediates the IL-4 signal and is essential for the proper development of adaptive immunity.
Host Species: Mouse
Clone: 18
Isotype: IgG2a
Species Reactivity: Human
Immunogen: Phosphorylated Human Stat6 (pY641)
Formula Weight [Chemical]: 100kDa
Immunofluorescence, Intracellular Staining, Western Blotting
Especificaciones
| Stat 6 (pY641) | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG2a | |
| Affinity Purified |
| 18/P-STAT6 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Phosphorylated Human Stat6 (pY641) | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.