missing translation for 'onlineSavingsMsg'
Learn More

SREBP-2 Mouse anti-Human, Unlabeled, Clone: IGG-1C6, BD

Código de producto. 15888818
Change view
Click to view available options
Cantidad:
0.1 mg
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15888818 0.1 mg cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15888818

Marca: BD Biosciences 557037

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

SREBP-1 and -2 (sterol-regulatory element binding proteins-1 and -2) are transcription factors which participate in the control of cholesterol homeostasis. SREBPs proteins, which are attached to the endoplasmic reticulum and nuclear envelope, are proteolytically cleaved and thus activated in response to conditions of low cellular sterol. Upon activation of SREBP-1 or -2, an ∽480-500 amino acid, N-terminal cleavage fragment of these proteins enters the nucleus and activates transcription of enzymes and other proteins required for cholesterol synthesis. Proteases which cleave SREBPs have been identified and include SCA (SREBP-cleavage activity), as well as a key regulator of apoptotic pathways, caspase-3. SREBP proteins containing point mutations at caspase-3 cleavage sites (Asp460 in SREBP-1 and Asp468 in SREBP-2) do not become cleaved following induction of apoptosis, suggesting that SREBPs may play some role in apoptotic processes. However, sterol-regulated vs. apoptosis-associated cleavage of SREBP proteins appears to be independantly regulated. On SDS-PAGE, sterol-regulated cleavage fragments of SREBP proteins migrate more slowly (i.e., higher molecular weight) than do staurosporin-induced fragments. In addition, staurosporin-induced SREBP cleavage products may appear as a doublet, with the upper band representing a phosphorylated form of SREBP. On SDS-PAGE, full length, precursor forms of SREBP-1 and -2 migrate at ∽125kDa, while proteolytic cleavage fragments may be observed as a cluster of bands between 60 - 70kDa. The IgG-2A4 antibody recognizes human and hamster SREBP-1. The antibody recognizes the N-terminal (basic helix-loop-helix) domain of human SREBP-1. A fusion protein containing N-terminal amino acids 301-407 (the bHLH/leucine zipper domain), was used as immunogen. The antibody recognizes both the 125kDa precursor and 60-70kDa mature, cleaved forms of SREBP-1.

Host Species: Mouse
Clone: IgG-1C6
Isotype: IgG1, κ
Species Reactivity: Human
Immunogen: Human SREBP-2 aa. 833-1141

Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno SREBP-2
Aplicaciones Western Blot
Clasificación Monoclonal
Clon IGG-1C6
Concentración 0.5mg/mL
Conjugado Unconjugated
Descripción Sterol Regulatory Element-Binding Factor 1; SREBF1
Formulación Aqueous buffered solution containing ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Human SREBP-2 aa. 833-1141
Método de purificación Affinity Purified
Cantidad 0.1 mg
Estado normativo RUO
Disciplina de investigación Cell Biology
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C
Isotype IgG1 κ
Mostrar más Mostrar menos

For Research Use Only.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.