Learn More
SNCG, Mouse, Clone: 2C3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SNCG.
Marca: Abnova H00006623-M01A.200uL
Descripción
Summary: This gene encodes a member of the synuclein family of proteins which are believed to be involved in the pathogenesis of neurodegenerative diseases. Mutations in this gene have also been associated with breast tumor development. [provided by RefSeq
Sequence: KTKQGVTEAAEKTKEGVMYVGAKTKENVVQSVTSVAEKTKEQANAVSEAVVSSVNTVATKTVEEAENIAVTSGVVRKEDLRPSAPQQEGEASKEKEEVAEEAQSGGDEspecificaciones
| SNCG | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC014098 | |
| SNCG | |
| SNCG (AAH14098, 21 a.a. ∼ 127 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6623 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 2C3 | |
| Mouse monoclonal antibody raised against a partial recombinant SNCG. | |
| SNCG | |
| BCSG1/SR | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.