missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sirtuin 1/SIRT1 Rabbit anti-Human, Mouse, Porcine, Amphibian, Bovine, Canine, Chicken, Chinese Hamster, Equine, Feline, Hamster, Primate, Rabbit, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
Marca: Novus Biologicals NBP2-27205
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Sirtuin 1/SIRT1 Polyclonal specifically detects Sirtuin 1/SIRT1 in Human, Mouse, Porcine, Amphibian, Bovine, Canine, Chicken, Chinese Hamster, Equine, Feline, Hamster, Primate, Rabbit samples. It is validated for Western Blot, Simple Western.Especificaciones
| Sirtuin 1/SIRT1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| SIRT1 | |
| A portion of amino acids 400-450 from human SIRT1 was used as the immunogen for this antibody. | |
| 0.1 mg | |
| Primary | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human, mouse, rat, orangutan, chimpanzee, panda, Carolina anole, cow, new world monkey, dog, Chinese hamster, horse, chicken, mole rat, rhesus monkey, wild turkey, opossum, platypus, rabbit, pig, zebra finch, frog, cat, and white-cheeked gibbon, and 89% homologous in zebrafish, cichlid fish, and killifish. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 1.0 mg/mL | |
| Western Blot 2-4ug/ml, Simple Western 1:100 | |
| EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 23411 | |
| Human, Mouse, Porcine, Amphibian, Bovine, Canine, Chicken, Chinese Hamster, Equine, Feline, Hamster, Primate, Rabbit | |
| Purified |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
For Research Use Only