Learn More
Siglec-1 (CD169) Mouse anti-Human, PE, Clone: 7-239, BD
Descripción
The 7-239 monoclonal antibody specifically binds to Sialic acid-binding Ig-like lectin 1 (Siglec-1), which is also known as Sialoadhesin (SN), or CD169. Siglec-1 is a type I transmembrane glycoprotein that belongs to the Siglec family within the Ig superfamily. This adhesion molecule especially binds to glycolipids and glycoproteins with terminal α-2 sialyl residues. Siglec-1 is expressed by macrophages and dendritic cells and serves as a cellular interaction molecule. Its expression can be upregulated by cells in response to type II collagen, or to cytokines including interferons, and tumor necrosis factor. Siglec-1 plays roles in endocytosis, hematopoiesis, and leucocyte migration. It mediates macrophage binding to various cell types including developing and mature leucocytes. Siglec-1 that is expressed by dendritic cells can also bind HIV-1 and may mediate viral transfer to bystander CD4+ T cells. Several Siglec-1 counter-receptors have been described including CD43, CD206, and CD227 which are expressed by T cells, macrophages, or breast cancer cells, respectively. The 7-239 antibody reportedly blocks Siglec-1 functions in some cellular assay systems.
Especificaciones
Especificaciones
| Antígeno | Siglec-1 (CD169) |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 7-239 |
| Conjugado | PE |
| Descripción | Sialoadhesin; SN; SIGLEC1; Siglec-1; Sialic acid-binding Ig-like lectin 1 |
| Formulación | Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
| Especie del huésped | Mouse |
| Inmunógeno | Human Rhinovirus-infected Dendritic Cells |
| Método de purificación | Affinity Purified |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.