Learn More
SERPINI1, Mouse, Clone: 1E10, Abnova™
Descripción
This gene encodes a member of the serpin superfamily of serine proteinase inhibitors. The protein is primarily secreted by axons in the brain, and preferentially reacts with and inhibits tissue-type plasminogen activator. It is thought to play a role in the regulation of axonal growth and the development of synaptic plasticity. Mutations in this gene result in familial encephalopathy with neuroserpin inclusion bodies (FENIB), which is a dominantly inherited form of familial encephalopathy and epilepsy characterized by the accumulation of mutant neuroserpin polymers. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | SERPINI1 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Monoclonal |
| Clon | 1E10 |
| Conjugado | Unconjugated |
| Descripción | Mouse monoclonal antibody raised against a full-length recombinant SERPINI1. |
| Formulación | PBS with no preservative; pH 7.4 |
| génica | SERPINI1 |
| N.º de referencia del gen | BC018043 |
| Alias de gen | DKFZp781N13156/PI12/neuroserpin |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.