Learn More
anti-S1P3/EDG-3, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-24762SS
Descripción
S1P3/EDG-3 Polyclonal specifically detects S1P3/EDG-3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Especificaciones
| S1P3/EDG-3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| EDG-3, EDG3FLJ37523, Endothelial differentiation G-protein coupled receptor 3, endothelial differentiation, sphingolipid G-protein-coupled receptor, 3, FLJ93220, G protein-coupled receptor, endothelial differentiation gene-3, LPB3, MGC71696, S1P receptor 3, S1P receptor Edg-3, S1P receptor EDG3, S1P3, sphingosine 1-phosphate receptor 3, Sphingosine 1-phosphate receptor Edg-3, sphingosine-1-phosphate receptor 3 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| 1903 | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1.0-2.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| Q99500 | |
| S1PR3 | |
| A portion of amino acids 300-350 of human Sphingolipid Receptor Edg3/S1P3 was used as the immunogen. | |
| 0.025 mg | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human and 95% homologous in mouse. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
For Research Use Only