Learn More
S100A4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA532576
Descripción
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is tonsil or melanoma tissue.
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.
Especificaciones
| S100A4 | |
| Polyclonal | |
| S100A4 | |
| 18A2, 42A, CAPL, FSP1, MTS1, P9KA, PEL98, S100 calcium-binding protein A4 calcium protein, calvasculin, metastasin, murine placental homolog, fibroblast-specific protein-1, leukemia multidrug resistance associated protein, maligt transformation suppression 1, placental calcium-binding protein, pr | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6275 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| P26447 | |
| S100A4 | |
| Recombit protein encoding full length human S100A4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.