Learn More
rhotekin, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant RTKN.
Marca: Abnova H00006242-A01.50uL
Descripción
This gene encodes a scaffold protein that interacts with GTP-bound Rho proteins. Binding of this protein inhibits the GTPase activity of Rho proteins. This protein may interfere with the conversion of active, GTP-bound Rho to the inactive GDP-bound form by RhoGAP. Rho proteins regulate many important cellular processes, including cytokinesis, transcription, smooth muscle contraction, cell growth and transformation. Dysregulation of the Rho signal transduction pathway has been implicated in many forms of cancer. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq
Sequence: VTDILTQREGARLETPPPWLAMFTDQPALPNPCSPASVAPAPDWTHPLPWGRPRTFSLDAVPPDHSPRARSVAPLPPQRSPRTRGLCSKGQPRTWLQSPEspecificaciones
| rhotekin | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant RTKN. | |
| RTKN | |
| RTKN | |
| RTKN (NP_149035, 451 a.a. ∼ 549 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 6242 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_033046 | |
| Mouse | |
| 50 μL | |
| Apoptosis | |
| Primary | |
| Human | |
| Serum |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.