missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF20 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human RNF20 protein.
Marca: Abnova H00056254-B01.50uL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The protein encoded by this gene shares similarity with BRE1 of S. cerevisiae. Yeast BRE1 is a ubiquitin ligase required for the ubiquitination of histone H2B and the methylation of histone H3. [provided by RefSeq
Sequence: MSGIGNKRAAGEPGTSMPPEKKAAVEDSGTTVETIKLGGVSSTEELDIRTLQTKNRKLAEMLDQRQAIEDELREHIEKLERRQATDDASLLIVNRYWSQFDENIRIILKRYDLEQGLGDLLTERKALVVPEPEPDSDSNQERKDDRERGEGQEPAFSFLATLASSSSEEMESQLQERVESSRRAVSQIVTVYDKLQEKVELLSRKLNSGDNLIVEEAVQELNSFLAQENMRLQELTDLLQEKHRTMSQEFSKLQSKVETAESRVSVLESMIDDLQWDIDKIRKREQRLNRHLAEVLERVNSKGYKVYGAGSSLYGGTITINARKFEEMNAELEENKELAQNRLCELEKLRQDFEEVTTQNEKLKVELRSAVEQVVKETPEYRCMQSQFSVLYNESLQLKAHLDEARTLLHGTRGTHQHQVELIERDEVSLHKKLRTEVIQLEDTLAQVRKEYEMLRIEFEQTLAANEQAGPINREMRHLISSLQNHNHQLKGEVLRYKRKLREAQSDLNKTRLRSGSALLQSQSSTEDPKDEPAELKPDSEDLSSQSSASKASQEDANEIKSKRDEEERERERREKEREREREREKEKEREREKQKLKESEKERDSAKDKEKGKHDDGRKKEAEIIKQLKIELKKAQESQKEMKLLLDMYRSAPKEQRDKVQLMAAEKKSKAELEDLRQRLKDLEDKEKKENKKMADEDALRKIRAVEEQIEYLQKKLAMAKQEEEALLSEMDVTGQAFEDMQEQNIRLMQQLREKDDANFKLMSERIKSNQIHKLLKEEKEELADQVLTLKTQVDAQLQVVRKLEEKEHLLQSNIGTGEKELGLRTQALEMNKRKAMEAAQLADDLKAQLELAQKKLHDFQDEIVENSVTKEKDMFNFKRAQEDISRLRRKLETTKKPDNVPKCDEILMEEIKDYKARLTCPCCNMRKKDAVLTKCFHVFCFECVKTRYDTRQRKCPKCNAAFGANDFHRIYIGEspecificaciones
| RNF20 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human RNF20 protein. | |
| RNF20 | |
| BRE1/BRE1A/FLJ11189/FLJ20382/KIAA2779/MGC129667/MGC129668/hBRE1 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 56254 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| No additive | |
| NM_019592.5 | |
| RNF20 | |
| RNF20 (NP_062538.5, 1 a.a. ∼ 975 a.a) full-length human protein. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido