missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-RIP3/RIPK3, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 23 publications
Marca: Novus Biologicals NBP2-24588SS
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
RIPK3/RIP3 Polyclonal specifically detects RIPK3/RIP3 in Human, Primate, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Especificaciones
| RIPK3/RIP3 | |
| Polyclonal | |
| Unconjugated | |
| 0.1 ml PBS with 0.05% Sodium Azide | |
| EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3 | |
| Rabbit | |
| 56.7 kDa | |
| 0.025 mg | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human and chimp. This antibody is not suitable for testing for mouse RIP3. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 3-7 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:20-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 10 ug/ml | |
| Q9Y572 | |
| RIPK3 | |
| RIPK3/RIP3 Antibody was made to a synthetic peptide (QEGPKDPEAWSRPQ) corresponding to amino acids 497-522 of human RIPK3/RIP3 was used as immunogen for this antibody (isoform CRA_a, 518 amino acids, NP_006862.2). This also corresponds to amino acids 298-311 of isoform CRA_b (GenBank: EAW66022.1). | |
| Affinity Purified | |
| RUO | |
| 11035 | |
| Human, Primate, Mouse (Negative) | |
| IgG |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
For Research Use Only