Learn More
anti-RHEB, Human, IP-WB Antibody Pair, Abnova™
Código de producto. 16078898
Change view
Click to view available options
Cantidad:
1 Set
Tamaño de la unidad:
1 set
16078898 1 Set 1 set
1 options
Código de producto. 16078898 Proveedor Abnova N.º de proveedor H00006009PW1.1Set
Includes
Antibody pair for IP: rabbit polyclonal anti-RHEB (300μL), Antibody pair for WB: mouse polyclonal anti-RHEB (50μL)
For immunoprecipitation and to detect the precipitated protein in western blot.
Especificaciones
Antígeno RHEB
Aplicaciones Immunoprecipitation, Western Blot
Clasificación Monoclonal/Polyclonal
Conjugado Unconjugated
Descripción This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
génica RHEB
Alias de gen MGC111559/RHEB2
Símbolos de los genes RHEB
Especie del huésped Mouse, Rabbit
Cantidad 1 Set
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6009
Incluye Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-RHEB (300 ul) 2. Antibody pair for WB: mouse polyclonal anti-RHEB (50 ul)
Especies diana Human
Contenido y almacenamiento Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Formulario Liquid
Mostrar más Mostrar menos
For Research Use Only