Learn More
renin binding protein, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant RENBP.
Marca: Abnova H00005973-A01.50uL
Descripción
The gene product inhibits renin activity by forming a dimer with renin, a complex known as high molecular weight renin. The encoded protein contains a leucine zipper domain, which is essential for its dimerization with renin. The gene product can catalyze the interconversion of N-acetylglucosamine to N-acetylmannosamine, indicating that it is a GlcNAc 2-epimerase. Transcript variants utilizing alternative promoters have been described in the literature. [provided by RefSeq
Sequence: FCPTQLEWAMKLWWPHSEAMIAFLMGYSDSGDPVLLRLFYQVAEYTFRQFRDPEYGEWFGYLSREGKVALSIKGGPFKGCFHVPRCLAMCEEMLGALLSREspecificaciones
| renin binding protein | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant RENBP. | |
| RENBP | |
| RBP/RNBP | |
| Mouse | |
| 50 μL | |
| Yes | |
| 5973 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_002910 | |
| RENBP | |
| RENBP (NP_002901, 301 a.a. ∼ 400 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.