missing translation for 'onlineSavingsMsg'
Learn More

Rb (pS780) Mouse anti-Human, Unlabeled, Clone: J146-35, BD

Código de producto. 15859338
Change view
Click to view available options
Cantidad:
0.1 mg
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15859338 0.1 mg cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15859338

Marca: BD Biosciences 558385

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The retinoblastoma gene product (Rb) is well known as a tumor suppressor and is either absent or mutated in many human tumors. Retrovirus-mediated gene transfer of the wild-type Rb gene into several Rb mutant neoplastic cell lines suppresses their tumorgenicity. Rb is a 110kDa nuclear phosphoprotein that undergoes differential phosphorylation during the cell cycle. During G1 phase, Rb is predominantly in a hypophosphorylated state. It becomes increasingly phosphorylated throughout the cell cycle until late mitosis, when substantial dephosphorylation occurs. Hypophosphorylated Rb interacts with a number of cellular proteins including the E2F transcription factor, several cyclins, RBP-1, RBP-2, c-Abl, c-myc, N-myc, and p46. Phosphorylation of Rb at various sites, by Cyclin-dependent protein kinases, inhibits the binding of Rb to these proteins. Rb is thought to mediate its effects, in part, via the repression of genes required for proliferation. For example, Rb is specifically recruited to promoters containing E2F sites and actively represses E2F mediated transcription. Rb also stimulates the activity of other transcription factors, although the mechanisms are less clearly defined. Thus, Rb appears to regulate transcription in its aim to control cell growth. The J146-35 monoclonal antibody recognizes Rb phosphorylated at serine 780 (pS780), which affects Rb binding to E2F. The orthologous phosphorylation sites in mouse and rat Rb are serines 773 and 751, respectively.

Immunohistochemistry, Western Blotting

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Rb (pS780)
Aplicaciones Western Blot
Clasificación Monoclonal
Clon J146-35
Concentración 0.5mg/mL
Conjugado Unconjugated
Formulación Aqueous buffered solution containing ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Phosphorylated Human Rb
Método de purificación Affinity Purified
Cantidad 0.1 mg
Estado normativo RUO
Disciplina de investigación Cell Biology
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.