Learn More
v-raf-1 murine leukemia viral oncogene homolog 1, Mouse, Clone: 1H4, Abnova™
Descripción
This gene is the cellular homolog of viral raf gene (v-raf). The encoded protein is a MAP kinase kinase kinase (MAP3K), which functions downstream of the Ras family of membrane associated GTPases to which it binds directly. Once activated, the cellular RAF1 protein can phosphorylate to activate the dual specificity protein kinases MEK1 and MEK2, which in turn phosphorylate to activate the serine/threonine specific protein kinases, ERK1 and ERK2. Activated ERKs are pleiotropic effectors of cell physiology and play an important role in the control of gene expression involved in the cell division cycle, apoptosis, cell differentiation and cell migration. Mutations in this gene are associated with Noonan syndrome 5 and LEOPARD syndrome 2. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | v-raf-1 murine leukemia viral oncogene homolog 1 |
| Aplicaciones | ELISA, In situ PLA, KnockDown, Western Blot |
| Clasificación | Monoclonal |
| Clon | 1H4 |
| Conjugado | Unconjugated |
| Descripción | Mouse monoclonal antibody raised against a partial recombinant RAF1. |
| Formulación | PBS with no preservative; pH 7.4 |
| génica | RAF1 |
| N.º de referencia del gen | BC018119 |
| Alias de gen | CRAF/NS5/Raf-1/c-Raf |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.