Learn More
RAB27 Mouse anti-Human, Unlabeled, Clone: 20.1/RAB27, BD
Descripción
Members of the Rab family of GTPases are non-transforming, membrane-bound, geranylgeranylated members of the Ras superfamily that are involved in the fusion and trafficking of cellular vesicles. Rab27A is expressed in many human tissues and it regulates the secreation of several types of cellular vesicles, such as melanosomes, insulin granules of pancreatic beta cells, and lytic granules of cytotoxic T lymphocytes. Rab27A is able to regulate such diverse processes by interacting with organelle-specific effector proteins, including melanophilin, granuphilin, and Munc13-4, respectively. Defects in Rab27A function are associated with the human diseases choroideremia and Griscelli syndrome 2 and the ashen mutant mouse. Rab27B is not so widely expressed, yet it is reported to have similar functional capacities as Rab27A. The 20.1 monoclonal antibody recognizes human Rab27A; cross-reactivity with Rab27B is unknown.
Western Blotting
Especificaciones
Especificaciones
| Antígeno | RAB27 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | 20.1/RAB27 |
| Concentración | 0.5mg/mL |
| Conjugado | Unconjugated |
| Formulación | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Human Rab27A Recombinant Protein |
| Método de purificación | Affinity Purified |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.