Learn More
PKA (RIIβ) (pS114) Mouse anti-Human, Alexa Fluor 647, Clone: 47, BD
Descripción
cAMP-dependent Protein Kinase (PKA) is composed of two distinct subunits: catalytic (C) and regulatory (R). Four regulatory subunits have been identified: RIα, RIβ, RIIα, and RIIβ. These subunits define type I and II PKAs. Following binding of cAMP, the regulatory subunits dissociate from the catalytic subunits, rendering the enzyme active. Type I and type II holoenzymes have three potential C subunits (Cα, Cβ, or Cγ). Most cells, including T lymphocytes, express both type I and type II PKAs. RIIa expression is associated with cellular transformation, while RIIβ expression correlates with mitotic arrest and cellular differentiation. Type II PKA can be distinguished by autophosphorylation of the R subunits, while type I PKA binds Mg/ATP with high affinity. The cAMP-dependent autophosphorylation of the human RIIβ subunits occurs at serine 114 (S114). In addition to their enzyme regulatory activity, the RIIα and RIIβ subunits determine the subcellular location of the holoenzymes via their interactions with specific intracellular anchoring proteins. The 47/PKA monoclonal antibody recognizes the phosphorylated S114 in the RIIβ subunit of PKA. The orthologous phosphorylation site in mouse and rat PKA(RIIβ) is S112.
Host Species: Mouse
Clone: 47
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Phosphorylated Human PKA(RIIβ) Peptide
Intracellular Staining
Especificaciones
Especificaciones
| Antígeno | PKA (RIIβ) (pS114) |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 47 |
| Conjugado | Alexa Fluor 647 |
| Descripción | KAP3, PKAR2B, PRKAR2B |
| Formulación | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Phosphorylated Human PKA(RIIβ) Peptide |
| Método de purificación | Affinity Purified |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.