Learn More
Phospho-IRF3 (Ser385) Polyclonal Antibody, Invitrogen™
Código de producto. 15570110
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
15570110 100 μg 100 microgramos
1 options
Código de producto. 15570110
Marca: Invitrogen PA538285
Rabbit Polyclonal Antibody
IRF-3 can inhibit cell growth and plays a critical role in controlling the expression of genes in the innate immune response. In unstimulated cells, IRF-3 is present in the cytoplasm. Viral infection results in phosphorylation of IRF-3 and leads to its translocation to the nucleus where it activates promoters containing IRF-3-binding sites. Phosphorylation of IRF-3 occurs at a cluster of C-terminal Ser and Thr residues (between 385 and 405), leading to its association with the p300/CBP coactivator protein that promotes DNA binding and transcriptional activity. During infection, IRF-3 is likely activated through a pathway that includes activation of Toll-like receptors and a kinase complex that includes IKK epsilon and TBK1. IRF-3 is phosphorylated at Ser396 following viral infection, expression of viral nucleocapsid, and double-stranded RNA treatment. These events likely play a role in activation of IRF-3.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Phospho-IRF3 (Ser385)
Aplicaciones Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
génica IRF3
N.º de referencia del gen Q14653, P70671
Alias de gen IRF3, IRF-3, IRF3, Interferon regulatory factor 3
Símbolos de los genes IRF3
Especie del huésped Rabbit
Inmunógeno A synthetic phosphopeptide derived from human IRF-3 around the phosphorylation site of Ser385 (G-A-SP-S-L)
Método de purificación Antigen affinity chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 292892, 3661, 54131
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG