Learn More
protein disulfide isomerase family A, member 3, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Descripción
This gene encodes a protein of the endoplasmic reticulum that interacts with lectin chaperones calreticulin and calnexin to modulate folding of newly synthesized glycoproteins. The protein was once thought to be a phospholipase; however, it has been demonstrated that the protein actually has protein disulfide isomerase activity. It is thought that complexes of lectins and this protein mediate protein folding by promoting formation of disulfide bonds in their glycoprotein substrates. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | protein disulfide isomerase family A, member 3 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Mouse polyclonal antibody raised against a full-length human PDIA3 protein. |
| Formulación | PBS with no preservative; pH 7.4 |
| génica | PDIA3 |
| N.º de referencia del gen | NM_005313.4 |
| Alias de gen | ER60/ERp57/ERp60/ERp61/GRP57/GRP58/HsT17083/P58/PI-PLC |
| Símbolos de los genes | PDIA3 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.