Learn More
PARP Mouse anti-Human, Unlabeled, Clone: 7D3-6, BD
Descripción
Includes
Purified Mouse Anti-Human PARP, Jurkat Cell Lysate
PARP [Poly(ADP-ribose) polymerase] is a 116kDa nuclear chromatin-associated enzyme that catalyzes the transfer of ADP-ribose units from NAD+ to a variety of nuclear proteins including topoisomerases, histones, and PARP itself. The catalytic activity of PARP is increased in nonapoptotic cells following DNA damage, and PARP is thought to play an important role in mediating the normal cellular response to DNA damage. Additionally, PARP is a target of the caspase protease activity associated with apoptosis. During apoptosis, PARP is cleaved from the 116kDa intact form into 85kDa and 25kDa fragments. This process separates the amino-terminal DNA-binding domain of the enzyme from the carboxy-terminal catalytic domain resulting in the loss of normal PARP function. Although the role of PARP in apoptosis remains to be elucidated, PARP cleavage is considered to be a marker of apoptosis.
Host Species: Mouse
Clone: 7D3-6
Isotype: IgG1
Species Reactivity: Human
Immunogen: Purified human PARP
Flow Cytometry, Immunoprecipitation, Western Blotting
Especificaciones
Especificaciones
| Antígeno | PARP |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | 7D3-6 |
| Concentración | 0.25mg/mL |
| Conjugado | Unconjugated |
| Especie del huésped | Mouse |
| Inmunógeno | Purified human PARP |
| Método de purificación | Affinity Purified |
| Cantidad | 150 μg |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.