Learn More
p54nrb Mouse, Unlabeled, Clone: 3, BD
Descripción
Splicing, the removal of introns from pre-mRNA, is mediated by spliceosomal complexes that contain multiple snRNPs and a number of non-snRNP splicing factors, such as hPRP17 and hPRP18. p54 [nrb] (nuclear RNA-binding protein, 54 kDa) is a non-snRNP RNA splicing factor. It binds and enhances the activity of topoisomerase I, the enzyme that nicks DNA to relieve torsional stress during replication and transcription. p54 [nrb] is the the human homolog of mouse NonO and bovine IPEB. The N-terminal half of p54 [nrb] contains two RNA recognition motifs (RRMs), that allow it to bind single stranded RNA. These motifs are included in a region that is 71% homologous with the human splicing factor PSF and 42% homologous with the Drosophila puff-specific protein BJ6. In addition to its role in RNA splicing, p54 [nrb] functions as a DNA transcription factor in multiple cell types. The N-terminal portion of p54 [nrb] also mediates its interaction with DNA. The ability of p54 [nrb] to bind DNA, but not RNA, is thought to require post-translational modification. Thus, p54 [nrb] can function as either an RNA splicing factor or a DNA transcription factor in a post-translational-dependent manner.
Especificaciones
Especificaciones
| Antígeno | p54nrb |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | 3 |
| Concentración | 250μg/mL |
| Conjugado | Unconjugated |
| Formulación | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Human p54 [nrb] aa. 368-471 |
| Método de purificación | Affinity Purified |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.