Learn More
Nkx6.1 Mouse, Unlabeled, Clone: R11-560, BD
Descripción
The R11-560 monoclonal antibody specifically binds to human and mouse homeobox protein Nkx6.1. The transcription factor Nkx6.1 plays a role in pancreatic islet beta cell development through the regulation of HNF1a and insulin genes. Nkx6.1 is considered to be a highly specific transcription factor marking pancreatic beta islet cells, where it is expressed in both developing and mature cells. In addition, Nkx6.1 functions as a transcriptional repressor induced by the Sonic Hedgehog (Shh) signaling pathway in fate determination of several neuronal cell types. During antibody development, the purified R11-560 monoclonal antibody was found to detect Nkx6.1 by western blot and indirect immunofluorescent staining followed by flow cytometric analysis or imaging analysis.
Especificaciones
Especificaciones
| Antígeno | Nkx6.1 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | R11-560 |
| Concentración | 0.5mg/mL |
| Conjugado | Unconjugated |
| Descripción | Nkx6.1, NKX6-1, NKX6A, Nkx6-1, Nkx6.1, Nkx6a |
| Formulación | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Human Nkx6.1 Recombinant Protein |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.