missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFkB p65 Rabbit anti-Human, Rabbit, Rat, Polyclonal, Invitrogen™
Descripción
PA5-16758 targets NFkappaB/p65 in IHC (P) and WB applications and shows reactivity with Human, Rabbit, and Rat samples. The PA5-16758 immunogen is a synthetic peptide corresponding to internal domain of human NFkappaB p65.
NFkappaB is a heterodimer consisting of a 50kDa DNA binding subunit and a 65kDa transactivating subunit (RelA). The p50/p65 heterodimer is present in the cytoplasm in inactive form bound to its inhibitor, IkappaB. In unstimulated cells, nuclear localization signals present on p65 are masked by members of the IkappaB family of inhibitory proteins. Subsequent to cell stimulation, IkappaB undergoes phosphorylation, ubiquitination and degradation by a proteosome-dependent pathway, allowing nuclear translocation of the active dimeric NFkappaB transcription factor.
Especificaciones
Especificaciones
| Antígeno | NFkB p65 |
| Aplicaciones | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA and 0.1% sodium azide; pH 7.6 |
| génica | RELA |
| N.º de referencia del gen | Q04206 |
| Alias de gen | nuclear factor NF-kappa-B p65 subunit; nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; transcription factor p65; v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor of kappa light polypeptide gene enhancer in B-cells 3 (p65)); v-rel reticuloendotheliosis viral oncogene homolog A; nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; p65; nf?b |
| Símbolos de los genes | RELA |
| Especie del huésped | Rabbit |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?