missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nestin Chicken anti-Human, Mouse, Rat, Baboon, Monkey, Polyclonal, Novus Biologicals™
Chicken Polyclonal Antibody has been used in 46 publications
Marca: Novus Biologicals NB100-1604
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Nestin Polyclonal specifically detects Nestin in Human, Mouse, Rat, Baboon, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Especificaciones
| Nestin | |
| Polyclonal | |
| Unconjugated | |
| 10 mM Sodium PBS (0.9% isotonic, w/v, pH 7.2) with 0.02% Sodium Azide | |
| FLJ21841, nestin | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| 10763 | |
| Human, Mouse, Rat, Baboon, Monkey | |
| IgY |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 0.1 mg/ml | |
| Western Blot 1:10000 - 1:20000, Flow Cytometry, Immunohistochemistry 1:1000 - 1:2000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:2000, Immunohistochemistry-Paraffin 1:1000 - 1:2000, Immunohistochemistry-Frozen 1:1000 - 1:2000 | |
| Q6P5H2, NP_057910 | |
| NES | |
| Chickens were immunized with three synthetic peptide/keyhole limpet hemocyanin (KLH) conjugates. These synthetic peptides corresponded to different regions of the mouse nestin gene product (NM_NP_057910), but are shared with the rat (NM_012987, NCBI) protein sequence. | |
| 0.25 mL | |
| Primary | |
| Nestin | |
| Store at 4C in the dark. |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
For Research Use Only