Learn More
Myogenin Mouse, Unlabeled, Clone: F5D, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 556358
Descripción
Myogenin is a member of the MyoD family of myogenic basic helix-loop-helix (bHLH) transcription factors that also includes MyoD, Myf-5, and MRF4 (also known as herculin or Myf-6). MyoD family members are expressed exclusively in skeletal muscle and play a key role in activating myogenesis by binding to enhancer sequences of muscle-specific genes. The regulatory domain of MyoD is approximately 70 amino acids in length and includes both a basic DNA binding motif and a bHLH dimerization motif. MyoD family members share about 80% amino acid homology in their bHLH motifs. Transfection of myogenin and other family members into a variety of non-muscle cells has been shown to either convert these cells to myogenic cells, or to transcriptionally activate a set of otherwise unexpressed muscle-specific genes. In addition to activating muscle-specific genes, members of the MyoD family members activate their own transcription and transactivate the transcription of other MyoD family members. For example, transfection of myogenin into 10T1/2 cells or Swiss 3T3 cells results in the activation of the endogenous myogenin gene as well as transactivation of MyoD. Likewise, the transfection of MyoD into these cells results in the activation of MyoD as well as the transactivation of myogenin. Each member of the MyoD family has distinct roles in muscle development; myogenin plays a key role in muscle maturation. Myogenin migrates at a molecular weight of ∽34kDa by SDS-PAGE. Clone F5D recognizes human, mouse, rat and cat myogenin. Mapping studies suggest that F5D reacts with an epitope between amino acids 138-158 of myogenin. A recombinant fusion protein corresponding to amino acids 30- 224 of rat myogenin was used as immunogen.
Host Species: Mouse
Clone: F5D
Isotype: IgG1
Species Reactivity: Human
Immunogen: Recombinant Rat Myogenin aa. 30-224
Immunohistochemistry, Immunohistochemistry (Frozen), Western Blotting
Especificaciones
| Myogenin | |
| Monoclonal | |
| 0.5mg/mL | |
| bHLHc3, Myf-4 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Western Blot | |
| F5D | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant Rat Myogenin aa. 30-224 | |
| 0.1 mg | |
| Cell Biology | |
| Feline, Human, Mouse, Rat | |
| IgG1 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only.