Learn More
MS4A8B, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human MS4A8B protein.
Marca: Abnova H00083661-B01P.50ug
Descripción
This gene encodes a member of the membrane-spanning 4A gene family. Members of this protein family are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues. The gene encoding this protein is localized to 11q12.3, among a cluster of family members. [provided by RefSeq
Sequence: MNSMTSAVPVANSVLVVAPHNGYPVTPGIMSHVPLYPNSQPQVHLVPGNPPSLVSNVNGQPVQKALKEGKTLGAIQIIIGLARIGLGSIMATVLVGEYLSISFYGGFPFWGGLWFIISGSLSVAAENQPYSYCLLSGSLGLNIVSAICSAVGVILFITDLSIPHPYAYPDYYPYAWGVNPGMAISGVLLVFCLLEFGIACASSHFGCQLVCCQSSNVSVIYPNIYAANPVITPEPVTSPPSYSSEIQANKEspecificaciones
| MS4A8B | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human MS4A8B protein. | |
| MS4A8B | |
| 4SPAN4/MS4A4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 83661 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC022895 | |
| MS4A8B | |
| MS4A8B (AAH22895, 1 a.a. ∼ 250 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.