missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MPS1 Monoclonal Antibody (7E3)

Descripción
MA5-15523 targets MPS1 in indirect ELISA, IF applications and shows reactivity with Human samples. The MA5-15523 immunogen is purified recombinant fragment of MPS1 expressed in E. Coli. MA5-15523 detects MPS1 which has a predicted molecular weight of approximately 9.5kDa.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S27E family of ribosomal proteins. It contains a C4-type zinc finger domain that can bind to zinc. The encoded protein has been shown to be able to bind to nucleic acid. It is located in the cytoplasm as a ribosomal component, but it has also been detected in the nucleus. Studies in rat indicate that ribosomal protein S27 is located near ribosomal protein S18 in the 40S subunit and is covalently linked to translation initiation factor eIF3. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Especificaciones
Especificaciones
| Antígeno | MPS1 |
| Aplicaciones | ELISA, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | 7E3 |
| Concentración | Conc. Not Determined |
| Conjugado | Unconjugated |
| Formulación | ascites with 0.03% sodium azide |
| génica | RPS27 |
| N.º de referencia del gen | P42677, Q6ZWU9 |
| Alias de gen | 3200001M24Rik; 40S ribosomal protein S27; metallopanstimulin 1; Metallopan-stimulin 1; MPS1; MPS-1; RGD:621045}; ribosomal protein S27; RP11-422P24.3; rps27; rps27 {ECO:0000312; S27; S27-1; Small ribosomal subunit protein eS27 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?