missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
The CD4 antibody from Proteintech is a rat monoclonal antibody generated with balb/c mouse thymocytes of mouse CD4. This antibody recognizes mouse antigen. The CD4 antibody has been validated for the following applications: FC analysis.
Especificaciones
Especificaciones
| Antígeno | CD4 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | RM4-4 |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Especie del huésped | Rat |
| Inmunógeno | BALB/c mouse thymocytes |
| Método de purificación | Protein A purification |
| Cantidad | 500 μg |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?