missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163 Mouse anti-Human, Porcine, Bovine, Guinea Pig, Primate, Monkey, Rhesus Macaque, Sheep, Clone: EDHu-1, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
Marca: Novus Biologicals NB110-40686
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD163 Monoclonal specifically detects CD163 in Human, Rat, Porcine, Bovine, Guinea Pig, Primate, Monkey, Rhesus Macaque, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay, Flow (Cell Surface).Especificaciones
| CD163 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay, Flow (Cell Surface) 1:10-1:1000 | |
| Q86VB7 | |
| CD163 | |
| Leucocytes harvested from pleural cavity of patients with idiopathic spontaneous pneumothorax. | |
| 0.1 mg | |
| Primary | |
| NB110-40686 recognizes the human CD163 cell surface antigen, a 130-140kD glycoprotein expressed by tissue macrophages. CD163 is not expressed by resting peripheral blood leucocytes, but expression may be induced on monocytes by culture in dexamethasone. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| EDHu-1 | |
| Unconjugated | |
| PBS with 0.09% Sodium Azide | |
| CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 9332 | |
| Human, Porcine, Bovine, Guinea Pig, Primate, Monkey, Rhesus Macaque, Sheep | |
| Purified |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
For Research Use Only