missing translation for 'onlineSavingsMsg'
Learn More

MMP3 Monoclonal Antibody, Clone: SPM293, Invitrogen™

MMP3 Antibody (SPM293), MA1-21234, from Invitrogen™. Species Reactivity: Human; Applications: IHC, Immunofluorescence, IHC (P), Western Blot.

Marca:  Invitrogen MA1-21234

Código de producto. 11563903

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos »

Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

MMP3 Antibody (SPM293), MA1-21234, from Invitrogen. Species Reactivity: Human; Applications: Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot.

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. MMP3 is an enzyme which degrades fibronectin, laminin, collagens III, IV, IX, and X, and cartilage proteoglycans. The enzyme is thought to be involved in wound repair, progression of atherosclerosis, and tumor initiation. MMP3 is reported to be implicated in rheumatoid arthritis and some forms of cancer.
Host Species: Mouse
Cross Reactivity: Human
Isotype: IgG2b
Clone: SPM293
Immunogen: APMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1).
Formulation: 250μL purified antibody (0.2mg/mL) in PBS with 1% BSA and sodium azide; pH 7.4
Synonyms: CHDS6, MGC126102, MGC126103, MGC126104, MMP-3, SL-1, STMY, STMY1, STR1
Uniprot Accession: P08254
Entrez GeneID: 4314
Gene Symbol: MMP3
MA1-21234 detects MMP-3 in human samples. MA1-21234 has been successfully used in immunofluorescence, immunohistochemistry (Paraffin) and Western blot procedures. Suggested positive controls include conditioned, serum-free medium from TPA treated human endometrial cells, placenta, bladder, breast, and ovarian carcinomas. MA1-21234 immunogen is APMA (4-Aminophenylmercuric acetate) activated human stromelysin-1 (SL-1).
TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

MMP3
Monoclonal
Unconjugated
CHDS6, MGC126102, MGC126103, MGC126104, MMP-3, SL-1, STMY, STMY1, STR1
Mouse
250 μL
Primary
Human
Purified
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
SPM293
P08254
MMP3
APMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1).
RUO
4314
Store at 4°C, do not freeze
IgG2b
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Promociones

Promociones

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado