missing translation for 'onlineSavingsMsg'
Learn More

MLH1 with control Mouse anti-Human, Unlabeled, Clone: G168-15, BD

Código de producto. 15886518
Change view
Click to view available options
Cantidad:
150 μg
50 μg
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15886518 150 μg cada uno
15876518 50 μg cada uno
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15886518

Marca: BD Biosciences 551092

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The repair of mismatched DNA is essential to maintaining the integrity of genetic information over time. In bacteria the DNA repair process is accomplished by the MutL, MutH, and MutS proteins. The MutS protein initially recognizes and binds to mismatched DNA. Following this, MutH, an endonuclease, and MutL form a complex with MutS and carry out an excision repair mechanism. When bacteria are deficient in one of these enzymes a mutator phenotype arises characterized by genetic instability. The important role played by DNA repair enzymes is emphasized by the fact that they are highly conserved from bacteria to yeast to mammals. In yeast the proteins are called MutS homolog 2 (MSH2), MutL homolog (MLH1), and PMS1 which is also a homolog of MutL. MSH2 is involved in the initial recognition of mismatched nucleotides during the replication mismatch repair process. It is thought that after MSH2 binds to a mismatched DNA duplex it is joined by a heterodimer of MLH1 and PMS1 which together help facilitate the later steps in mismatch repair. Biochemical studies of the human homologs of DNA mismatch repair enzymes MLH1, PMS2, and MSH2 indicate that human MSH2 protein can bind mispaired DNA, and that human MLH1 and PMS2 can exist as a heterodimer. These and other studies support the conservation of eukaryotic DNA mismatch repair mechanisms. The G168-15 antibody recognizes human MLH1 (80-85 kDa). Full-length human recombinant MLH was expressed as a fusion protein, affinity purified, and used as immunogen.

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno MLH1 with control
Aplicaciones Western Blot
Clasificación Monoclonal
Clon G168-15
Conjugado Unconjugated
Especie del huésped Mouse
Método de purificación Affinity Purified
Cantidad 150 μg
Estado normativo RUO
Primario o secundario Primary
Incluye Purified Mouse Anti-Human MLH-1 (150μg) 3 vials of 50μg; Jurkat Cell Lysate (50μg)
Especies diana Human
Contenido y almacenamiento Store undiluted at -20°C.
Isotype IgG1
Mostrar más Mostrar menos

For Research Use Only.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.