Learn More
Mitosin Mouse anti-Human, Unlabeled, Clone: 11, BD
Descripción
Mitosin, a nuclear proetin of 3113 amino acids, contains a pair of tandem repeats and seven leucine repeats. In vitro, Mitosin directly interacts with the retinoblastoma protein, Rb. Mitosin is expressed throughout the cell cycle, with levels being lowest during G1, and it localizes at the kinetochore during the mitotic phase. The subcellular redistribution of Mitosin to the kinetochore is linked to its phosphorylation. Its expression is linked to patients with autoimmune diseases characterized by abnormal cell proliferation. Ectopic expression of a truncated version of Mitosin blocks the progression of the cell cycle. This suggests that the protein has an important role during cell proliferation. Because of similarities in subcellular localization and expression, Mitosin and the autoantigen p300/CENP-F, which is also linked to a multitude of autoimmune disorders, are probably the same protein.
Immunofluorescence, Immunoprecipitation, Western Blotting
Especificaciones
Especificaciones
| Antígeno | Mitosin |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | 11 |
| Concentración | 250μg/mL |
| Conjugado | Unconjugated |
| Formulación | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Human Mitosin aa. 209-381 |
| Método de purificación | Affinity Purified |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.