missing translation for 'onlineSavingsMsg'
Learn More

MITF Polyclonal Antibody, Invitrogen™

Código de producto. 11526682
Change view
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
50 microgramos
Código de producto. Cantidad unitSize
11526682 50 μg 50 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11526682

Marca: Invitrogen PA127613

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

MITF Antibody, PA1-27613, from Invitrogen™. Species Reactivity: Human; Applications: Western Blot.

MITF Antibody, PA1-27613, from Invitrogen. Species Reactivity: Human; Applications: Western Blot.

Transcription factor for tyrosinase and tyrosinase-related protein 1. Binds to a symmetrical DNA sequence (E-boxes) (5'-CACGTG-3') found in the tyrosinase promoter. Plays a critical role in the differentiation of various cell types as neural crest-derived melanocytes, mast cells, osteoclasts and optic cup-derived retinal pigmant epithelium. This protein is a transcription factor that contains both basic helix-loop-helix and leucine zipper structural features. It regulates the differentiation and development of melanocytes retinal pigment epithelium and is also responsible for pigment cell specific transcription of the melanogenesis enzyme genes. Heterozygous mutations in the this gene cause auditory-pigmentary syndromes, such as Waardenburg syndrom type 2 and Tietz syndrome. Alternateively spliced transcript variants encoding different isoforms have been identified.
Host Species: Chicken
Cross Reactivity: Human
Isotype: IgY
Immunogen: Synthetic peptide corresponding to human Microphthalmia-associated transcription factor.
Formulation: 50μg purified antibody in PBS and no preservative
Synonyms: MI, WS2A, bHLHe32
Uniprot Accession: O75030
Entrez GeneID: 4286
Gene Symbol: MITF
PA1-27613 detects MITF from human samples. PA1-27613 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein. The immunogen is a synthetic peptide corresponding to human Microphthalmia-associated transcription factor. Store at 4ºC short term. For extended storage aliquot and store at -20ºC or below. Avoid freeze-thaw cycles.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno MITF
Aplicaciones Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
N.º de referencia del gen O75030
Alias de gen MI, WS2A, bHLHe32
Símbolos de los genes MITF
Especie del huésped Chicken
Inmunógeno Synthetic peptide corresponding to human Microphthalmia-associated transcription factor.
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 4286
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Formulario Purified
Isotype IgY
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.