missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP2 Polyclonal Antibody
Descripción
It is not recommended to aliquot this antibody. PA5-17646 was successfully used to detect MAP2 in neurons differentiated from H9 ESC derived NSCs. This antibody is not cross-reactive with tau.
MAP2 (Microtubule Associated Protein 2) exists in two high molecular weight forms (MAP2a & MAP2b) and a low molecular weight form. The expression of MAP2 is developmentally regulated and its multiple forms arise by alternative splicing of a single gene. MAP2 is involved in microtubule assembly, which is an essential step in neurogenesis. The products of similar genes in rat and mouse are neuron-specific cytoskeletal proteins that are enriched in dendrites, implicating a role in determining and stabilizing dendritic shape during neuron development. MAP2 is useful in studies of neuron structure in normal and malignant brain tissue as well as in degenerative diseases such as Alzheimer's Disease (AD).
Especificaciones
Especificaciones
| Antígeno | MAP2 |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 73.6 μg/mL |
| Conjugado | Unconjugated |
| Formulación | 0.01M HEPES with 100μg/mL BSA, 0.15M NaCl, 50% glycerol and no preservative; pH 7.5 |
| génica | Map2 |
| N.º de referencia del gen | P11137, P15146, P20357 |
| Alias de gen | a730034c02; AC091465.1; DKFZp686I2148; G1-397-34; MAP 2; Map2; MAP-2; map2.S; map2a; map-2a; map2ab; MAP2B; MAP2C; MAP2R; microtubule associated protein 2; microtubule associated protein 2 S homeolog; microtubule associated protein 2; microtubule-associated protein 2; microtubule associated protein 2C; microtubule-associated protein 2; microtubule-associated protein-2; Mtap2; Mtap-2; OTTHUMP00000163916; OTTHUMP00000208143; repro4; XELAEV_18047318mg; xmap2 |
| Símbolos de los genes | Map2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?