missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-M-CSF Receptor/CD115 FITC, Clone: AFS98, Invitrogen™
Descripción
M-CSF Receptor/CD115 Monoclonal antibody A27078 from Invitrogen specifically detects M-CSF Receptor/CD115 in Mouse samples. It is validated for Flow Cytometry.
CSF1R is the receptor for colony stimulating factor 1, a cytokine which controls the production, differentiation, and function of macrophages. This receptor mediates most if not all of the biological effects of this cytokine. Ligand binding activates the receptor kinase through a process of oligomerization and transphosphorylation. The encoded protein is a tyrosine kinase transmembrane receptor and member of the CSF1/PDGF receptor family of tyrosine-protein kinases.
Host Species: Rat
Cross Reactivity: Mouse
Isotype: IgG2a, κ
Clone: AFS98
Immunogen: Mouse CD115
Formulation: 100μg Purified Antibody in PBS, with 0.1% gelatin and 0.09% sodium azide
Synonyms: CSF1R, colony stimulating factor 1 receptor, C-FMS, CD115, CSFR, FIM2, FMS, CD115 antigen, FMS proto-oncogene, McDonough feline sarcoma viral v-fms oncogene homolog, macrophage colony stimulating factor I receptor
Uniprot Accession: P09581
Entrez GeneID: 12978
Gene Symbol: Csf1r
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
Host Species: Rat
Cross Reactivity: Mouse
Isotype: IgG2a, κ
Clone: AFS98
Immunogen: Mouse CD115
Formulation: 100μg Purified Antibody in PBS, with 0.1% gelatin and 0.09% sodium azide
Synonyms: CSF1R, colony stimulating factor 1 receptor, C-FMS, CD115, CSFR, FIM2, FMS, CD115 antigen, FMS proto-oncogene, McDonough feline sarcoma viral v-fms oncogene homolog, macrophage colony stimulating factor I receptor
Uniprot Accession: P09581
Entrez GeneID: 12978
Gene Symbol: Csf1r
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
Especificaciones
Especificaciones
| Antígeno | M-CSF Receptor/CD115 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | AFS98 |
| Conjugado | FITC |
| Formulación | 100μg Purified Antibody in PBS, with 0.1% gelatin and 0.09% sodium azide |
| N.º de referencia del gen | P09581 |
| Alias de gen | CSF1R, colony stimulating factor 1 receptor, C-FMS, CD115, CSFR, FIM2, FMS, CD115 antigen, FMS proto-oncogene, McDonough feline sarcoma viral v-fms oncogene homolog, macrophage colony stimulating factor I receptor |
| Símbolos de los genes | Csf1r |
| Especie del huésped | Rat |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?