missing translation for 'onlineSavingsMsg'
Learn More

Lck (pY505) Mouse, Alexa Fluor 647, Clone: 4/LCK-Y505, BD

Código de producto. 13470059
Change view
Click to view available options
Cantidad:
50 Tests
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
13470059 50 Tests cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 13470059

Marca: BD Biosciences 558577

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Lck is a member of the Src family of cytoplasmic protein-tyrosine kinases (PTKs) that is normally expressed exclusively in lymphoid cells, primarily T lymphocytes and NK cells. Members of this family have several common features: 1) unique N-terminal domains, 2) attachment to cellular membranes through a myristylated N-terminus, and 3) homologous SH2, SH3, and catalytic domains. The unique N-terminal domain of Lck interacts with the cytoplasmic tails of the CD4 and CD8 cell-surface glycoproteins of T lymphocytes, which recognize antigen presenting cells via their surface MHC class II and class I molecules, respectively. The catalytic activity of Lck is regulated by both kinases and phosphatases that control the phosphorylation states of two tyrosine residues that have opposing effects. Repression of Lck's catalytic activity occurs via phosphorylation at tyrosine 505 (Y505), located near the carboxy terminus. Phosphorylation of this tyrosine site is mediated by the Csk family of PTKs, and its dephosphorylation is mediated by the protein tyrosine phosphatase, CD45. When Lck is phosphorylated at this site, it assumes a folded tertiary structure which is enzymatically inactive. When CD45 dephosphorylates it at Y505, Lck is able to autophosphorylate its Y394, which leads to conformational changes in the catalytic domain that induce kinase activity. However, it has been observed that the inhibitory effect of the phosphorylated Y505 can be overcome by direct engagement of Lck's SH3 domain and that both Y394 and Y505 are phosphorylated together in cells activated by hydrogen peroxide. Activated Lck phosphorylates the ITAMs (Immunoreceptor-based Tyrosine Activation Motifs) of the T cell receptor (TCR) and thus is critical for activation and development of T lymphocytes. The interactions of Lck, Csk, CD45, CD4 or CD8, and TCR are only a small part of a complex immunoregulatory cascade that involves additional substrates for Csk and CD45, other enzymes, adhesion molecules, adaptor proteins, and specialized membrane microdomains. The 4/LCK-Y505 monoclonal antibody recognizes the phosphorylated Y505 of the catalytic domain of Lck.The Alexa Fluor™ 488- conjugated format has been evaluated by flow using a human model system. However, the unconjugated form of this antibody (Mfr. No. 612390) has been shown to react with human, mouse, and rat in western blot. A phosphorylated peptide corresponding to residues around Tyrosine-505 from human Lck was used as the immunogen.

Intracellular Staining

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Lck (pY505)
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 4/LCK-Y505
Conjugado Alexa Fluor 647
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Phosphorylated Human Lck Peptide
Método de purificación Affinity Purified
Cantidad 50 Tests
Estado normativo RUO
Disciplina de investigación Cell Biology
Primario o secundario Primary
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.