Learn More
Langerin (CD207) Mouse anti-Human, PE, Clone: 2G3 (also known as AB5_8.2G3.6 ), BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 564727
Descripción
The 2G3 monoclonal antibody specifically binds to Langerin which is also known as CD207 or C-type lectin domain family 4 member K (CLEC4K). CD207 is a 48 kDa type II transmembrane glycoprotein that belongs to the C-type lectin family. CD207 is selectively expressed by Langerhans cells, an immature dendritic cell type that is found in the epidermis and mucosal epithelia, and by some other dendritic cell subsets. CD207 is expressed on the cell surface membrane and by intracellular Birbeck granules whose formation depends on CD207. CD207 binds to certain surface carbohydrates that are expressed by microbes. It appears to play a role in the uptake of these microbes for subsequent processing and microbial antigen presentation to T cells.
Especificaciones
| Langerin (CD207) | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human Langerin Recombinant Protein | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 2G3 (also known as AB5_8.2G3.6 ) | |
| CLC4K; CLEC4K; C-type lectin domain family 4 member K | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 λ |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.