missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KISS1R Polyclonal Antibody

Descripción
Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey (95%) Marmoset (80%).
GCPR54 is a galanin-like G protein coupled receptor that binds metastin, a peptide encoded by the metastasis suppressor gene KISS1. The tissue distribution of the protein suggests that it is involved in the regulation of endocrine function, and this is supported by the finding that it appears to play a role in the onset of puberty. GPR54 has been reported in various human brain regions, placenta, pancreas, and spinal cord, and at lower levels in spleen, peripheral blood leukocytes, testis, lymph node, thymus, small intestine, stomach, lung, and kidney. The receptor is also abundant in tumor tissues. In rat, GPR54 is expressed in liver, intestine, and in the pons, midbrain, thalamus, hypothalamus, hippocampus, amygdala, cortex (including frontal), and striatum. ESTs have been isolated from normal placenta and kidney cancer libraries.
Especificaciones
Especificaciones
| Antígeno | KISS1R |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.1% sodium azide |
| génica | KISS1R |
| N.º de referencia del gen | Q969F8 |
| Alias de gen | AXOR12; CPPB1; G protein-coupled receptor 54; Gpr54; G-protein coupled receptor 54; G-protein coupled receptor OT7T175; HH8; HOT7T175; Hypogonadotropin-1; kiSS-; KiSS-1; KISS1 receptor; kiSS-1 receptor; KISS1R; KiSS-1R; Kisspeptins receptor; metastin receptor; mOT7T175; orphan G protein-coupled receptor 54; orphan G protein-coupled receptor GPR54; rOT7T175 |
| Símbolos de los genes | KISS1R |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?