Learn More
KIF3A Mouse, Unlabeled, Clone: 28, BD
Descripción
The ability of the kinesin superfamily of motor proteins to hydrolyze ATP as they move progressively along microtubules is important for organelle transport and cell division. Kinesins are grouped according to the location of the motor domain in the N-terminal, middle, or C-terminal region of the protein. A family of N-terminal motor domain kinesin proteins includes KIF3A/3B, KRP85/95, and Klp68d/64. These proteins form heterodimeric two-headed motors that interact with kinesin associated protein (KAP). The KIF3A/3B/KAP3 heterotrimeric motor has plus-end directed microtubule sliding activity. This heterotrimeric motor is expressed ubiquitously and is used for anterograde transport of membranous organelles. KIF3A- and KIF3B-deficient mice have defects in the formation of the cilia located in the embryonic node. In addition, KIF3A localizes to the basal body of the connecting cilium axoneme and to vesicles at the pre-synaptic ribbon in photoreceptor cells. Thus, KIF3 motors are involved in microtubule-based formation and movement of cilia and the transport of synaptic vesicles.
Host Species: Mouse
Clone: 28
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Human KIF3A aa. 563-671
Formula Weight [Chemical]: 80/85kDa
Immunofluorescence, Western Blotting
Especificaciones
Especificaciones
| Antígeno | KIF3A |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | 28 |
| Concentración | 250μg/mL |
| Conjugado | Unconjugated |
| Formulación | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Human KIF3A aa. 563-671 |
| Método de purificación | Affinity Purified |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.