Learn More
IL-6 Rat anti-Human, PE-CF594, Clone: MQ2-13A5, BD
Descripción
The MQ2-13A5 monoclonal antibody specifically binds to human interleukin-6 (IL-6). IL-6 is a multifunctional cytokine that plays a central role in host defense mechanisms, including hematopoiesis, immune responses (eg, T and B cell activation and differentiation) and acute phase reactions. IL-6 can be expressed by a variety of cells including monocytes/macrophages, eosinophils, fibroblasts, vascular endothelial cells, bone marrow stromal cells, mesangial cells, hepatocytes, keratinocytes, astrocytes, T lymphocytes, B lymphocytes, and various tumor cells. IL-6 production is upregulated by cells in response to bacterial products such as lipopolysaccharide, viruses and other pro-inflammatory cytokines such as IL-1, TNF , and IFN-γ. IL-6 transcription is downregulated by cells in response to IL-4 and IL-10. The functional IL-6 Receptor (IL-6R) complex consists of two transmembrane glycoproteins, an 80-kDa low-affinity ligand-binding receptor subunit (IL-6Rα/CD126) and a 130 kDa (gp130/CD130) subunit that binds to IL-6-IL-6Rα to form the high-affinity signal transducing complex. Abnormal expression of IL-6 is related to the pathogenesis of many diseases including neoplastic (eg, multiple myeloma) and autoimmune diseases (eg, rheumatoid arthritis). The immunogen used to generate this hybridoma was COS-7 -expressed recombinant human IL-6.
Especificaciones
Especificaciones
| Antígeno | IL-6 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | MQ2-13A5 |
| Conjugado | PE-CF594 |
| Descripción | IL6; Interleukin-6; BSF-2; CDF; HGF; HSF; IFNB2 |
| Formulación | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Especie del huésped | Rat |
| Inmunógeno | Human IL-6 Recombinant Protein |
| Método de purificación | Affinity Purified |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.