missing translation for 'onlineSavingsMsg'
Learn More

IL-21 Mouse anti-Human, PE, Clone: 3A3-N2.1 (also known as 3A3-N2), BD

Código de producto. 15851589
Change view
Click to view available options
Cantidad:
25 Tests
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15851589 25 Tests cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15851589

Marca: BD Biosciences 562042

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Human Interleukin-21 (IL-21) is a member of the type I cytokine family that is encoded by a gene resident on chromosome 4. The mature form of human IL-21 is a 131 amino acid protein. IL-21 is produced by activated NKT and multiple CD4+ T cell subsets including effector memory and central memory CD4+ T cells and differentiated T helper cell subsets polarized towards Th17 cell and T follicular helper (Tfh) phenotypes. IL-21 plays important protective roles in the regulation of hematopoiesis and both innate and adaptive immune responses and adverse roles in promoting autoimmunity. IL-21 costimulates the proliferation and differentiation of CD4+ T cells. It enhances the proliferation of and cytotoxicity mediated by natural killer (NK) cells and CD8+ T cells. IL-21 costimulates B cell proliferation and differentiation into plasma cells producing immunoglobulins with IgG isotypes. IL-21 can also regulate the functions of dendritic cells and other myeloid cells. IL-21 exerts its biological activities by binding to and activating the Janus activating kinases (JAK1 and JAK3) and signal transducers and activators of transcription (STAT1, STAT3, STA5a and STAT5b) signaling pathways through the IL-21 receptor (IL-21R) complex. The IL-21R complex is comprised of the IL-21R alpha subunit and the common cytokine receptor gamma subunit (γ c; CD132).

The monoclonal 3A3-N2 antibody specifically binds to human IL-21.

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno IL-21
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 3A3-N2.1 (also known as 3A3-N2)
Conjugado PE
Descripción Interleukin-21, IL21, Za11
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Human IL-21 Recombinant Protein
Método de purificación Affinity Purified
Cantidad 25 Tests
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.