Learn More
IL-10 Rat anti-Human and Viral, Brilliant Violet 711, Clone: JES3-9D7, BD
Descripción
The JES3-9D7 monoclonal antibody specifically reacts with human IL-10 (Interleukin-10) and viral IL-10. The immunogen used to generate the JES3-9D7 hybridoma was recombinant human IL-10 expressed by COS cells. IL-10 is also known as CSIF (Cytokine synthesis inhibitory factor) and (TGIF) T-cell growth inhibitory factor. IL-10 is expressed by various cell types including activated monocytes, macrophages, dendritic cells, mast cells, granulocytes, and lymphocytes. IL-10 is a pleiotropic cytokine that can downregulate proinflammatory immune responses, such as Th1-like responses, while promoting other responses including B cell proliferation and antibody production.
The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon™ BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy™5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.
Especificaciones
Especificaciones
| Antígeno | IL-10 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | JES3-9D7 |
| Conjugado | Brilliant Violet 711 |
| Descripción | CSIF; Cytokine synthesis inhibitory factor; TGIF |
| Formulación | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Especie del huésped | Rat |
| Inmunógeno | Recombinant Human IL-10 |
| Método de purificación | Affinity Purified |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.