Learn More
anti-IgM, κ, APC, Clone: G155-228, BD
Marca:  BD Biosciences 555585
Código de producto. 15848288
  • 127.00€ / cada uno
Descripción
This antibody reacts with a 45-48kDa type I integral membrane glycoprotein present on peripheral blood and tonsillar B cells, but not expressed on terminally differentiated B cells. CD40 is also present on some epithelial cells, carcinomas and lymphoid dendritic cells. This antibody is useful for studies of B-cell growth and function. Anti-CD40 antibodies have been reported to stimulated B-cell proliferation when costimulated with anti-?, anti-CD20 antibodies or with phorbol esters. 5C3 is capable of inducing B-cell proliferation when presented with IL-4. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only.
Flow Cytometry
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
Flow Cytometry
APC
Affinity Purified
Undiluted at 4°C. Protect from exposure to light. Do not freeze.
G155-228
Mouse
100 Tests
IgM κ