Learn More
anti-IgG1, PE, Clone: X40, BD
Descripción
The Mouse IgG1 antibody, clone X40, is derived from the hybridization of Sp2/0 mouse myeloma cells with spleen cells isolated from BALB/c mice immunized with KLH. The Mouse IgG1 antibody is specific for keyhole limpet hemocyanin (KLH), an antigen not expressed on human peripheral blood platelets.
Especificaciones
Especificaciones
| Antígeno | IgG1 |
| Clasificación | Monoclonal |
| Clon | X40 |
| Conjugado | PE |
| Cantidad | 50 μg |
| Estado normativo | RUO |
| Disciplina de investigación | Immunology |
| Primario o secundario | Primary |
| Especies diana | Human |